1-(2,4,6-Trimethylphenyl)-2-(dicyclohexylphosphino)imidazole ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: cataCXium® PlCy

15-6362-500MG 15-6362-2G
100198-670EA 157.11 USD
100198-670 100206-424
1-(2,4,6-Trimethylphenyl)-2-(dicyclohexylphosphino)imidazole ≥95%
1-(2,4,6-Trimethylphenyl)-2-(dicyclohexylphosphino)imidazole

cataCXium, Phosphine

Melting Pt: 109 °C
MDL Number: MFCD06658125
CAS Number: 794527-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR