2,4,4-Trimethylpentylphosphine ≥99% (8% isomers)

Supplier: STREM CHEMICALS INC MS
Danger

15-6360-25G 15-6360-5G
100199-224EA 1113.91 USD
100199-224 100199-226
2,4,4-Trimethylpentylphosphine ≥99% (8% isomers)
2,4,4-Trimethylpentylphosphine

Phosphine

Flash Pt: 1 °F
MDL Number: MFCD00269836
CAS Number: 82164-75-8
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR