Triethylphosphine oxide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-6310-5G 15-6310-1G
100199-808EA 435.94 USD
100199-808 100206-418
Triethylphosphine oxide ≥98%
Triethylphosphine oxide
Formula: C₆H₁₅OP
MW: 134.16 g/mol
Boiling Pt: 84…85 °C (3 mmHg)
Melting Pt: 52…53 °C
MDL Number: MFCD00013914
CAS Number: 597-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR