Tri-tert-butylphosphonium tetrafluoroborate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tri-t-butylphosphonium tetrafluoroborate
15-6000-1G
15-6000-5G
100204-974EA
106.15
USD
100204-974
100204-976
Tri-tert-butylphosphonium tetrafluoroborate ≥99%
Tri-tert-butylphosphonium tetrafluoroborate
Phosphine
Formula:
C₁₂H₂₈BF₄P MW: 290.13 g/mol Melting Pt: 261 °C (dec.) |
MDL Number:
MFCD04039975 CAS Number: 131274-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...