Tributylphosphonium tetrafluoroborate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tri-n-butylphosphonium tetrafluoroborate

15-5990-5G 15-5990-1G
100209-734EA 202.4 USD
100209-734 100209-732
Tributylphosphonium tetrafluoroborate ≥99%
Tributylphosphonium tetrafluoroborate
Formula: C₁₂H₂₈BF₄P
MW: 290.13 g/mol
Melting Pt: 51…52 °C
MDL Number: MFCD05664257
CAS Number: 113978-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR