Tributyl phosphite ≥94%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tri-n-butylphosphite

15-5950-250G 15-5950-1000G
100198-666EA 90.58 USD
100198-666 100207-990
Tributyl phosphite ≥94%
Tributyl phosphite
Formula: C₁₂H₂₇O₃P
MW: 250.32 g/mol
Boiling Pt: 121…125 °C (6 mmHg)
Density: 0,925
Flash Pt: 197 °F
MDL Number: MFCD00009437
CAS Number: 102-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR