(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-4,4'-Bis(bis(3,5-dimethylphenyl)phosphino)-2,2',6,6'-tetramethoxy-3,3'-bipyridine, CTH-(R)-Xylyl-P-PHOS

15-5210-100MG 15-5210-500MG
100204-966EA 107.57 USD
100204-966 100204-968
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine ≥97%
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine

Phosphine, P-PHOS

Formula: C₄₆H₅₀N₂O₄P₂
MW: 756.85 g/mol
MDL Number: MFCD04974235
CAS Number: 442905-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR