(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (11aS)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine, (S)-SIPHOS-PE

15-5163-500MG 15-5163-100MG
101418-690EA 904.43 USD
101418-690 101418-688
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine ≥98%
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine

MONOPHOS, Phosphine, SIPHOS

Formula: C₃₃H₃₂NO₂P
MW: 505.58 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD08459342
CAS Number: 500997-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR