(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-SIPHOS-PE, (11aR)-10,11,12,13-Tetrahydro-N,N-bis[(1R)-1-phenylethyl]diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-amine

15-5162-500MG 15-5162-100MG
101418-686EA 914.34 USD
101418-686 101418-684
(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine ≥98%
(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine

MONOPHOS, Phosphine, SIPHOS

Formula: C₃₃H₃₂NO₂P
Melting Pt: 98…100 °C
MDL Number: MFCD08459342
CAS Number: 500997-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR