(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-5150-500MG 15-5150-100MG
101418-670EA 733.17 USD
101418-670 101418-668
(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%
(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine

MONOPHOS, Phosphine, SIPHOS

Formula: C₁₉H₂₀NO₂P
Melting Pt: 95…96 °C
MDL Number: MFCD08459340
CAS Number: 443965-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR