Dimethyl phenylphosphonite ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Phenyldimethoxyphosphine

15-4250-5G 15-4250-1G
100209-726EA 142.95 USD
100209-726 100204-956
Dimethyl phenylphosphonite ≥98%
Dimethyl phenylphosphonite

Phosphine

Formula: C₈H₁₁O₂P
MW: 170.15 g/mol
Boiling Pt: 77…79 °C (7 mmHg)
Density: 1,072
Flash Pt: >230 °F
MDL Number: MFCD00008352
CAS Number: 2946-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR