Phenylphosphonic dichloride ≥94%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide

15-4158-250G 15-4158-50G
100207-986EA 407.63 USD
100207-986 100196-098
Phenylphosphonic dichloride ≥94%
Phenylphosphonic dichloride
Formula: C₆H₅Cl₂OP
MW: 194.98 g/mol
Boiling Pt: 258 °C
Melting Pt: 3 °C
Density: 1.394 (25°)
Flash Pt: 400 °F
MDL Number: MFCD00002070
CAS Number: 824-72-6
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR