N-Phenyl-2-(di-t-butylphosphino)pyrrole ≥95%

Supplier: STREM CHEMICALS INC MS

Synonyms: 2-(di-tert-Butylphosphino)-1-phenyl-1H-pyrrole, cataCXium® PtB

15-3600-2G 15-3600-500MG
100206-394EA 460 USD
100206-394 100206-392
N-Phenyl-2-(di-t-butylphosphino)pyrrole ≥95%
N-Phenyl-2-(di-t-butylphosphino)pyrrole

cataCXium, Phosphine

Formula: C₁₈H₂₆NP
MW: 287.379 g/mol
Melting Pt: 51 °C
MDL Number: MFCD05861612
CAS Number: 672937-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR