N-Phenyl-2-(di-t-butylphosphino)indol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-(Di-tert-butylphosphino)-1-phenyl-1H-indole, cataCXium® PIntB

15-3550-2G 15-3550-500MG
100206-390EA 460 USD
100206-390 100198-650
N-Phenyl-2-(di-t-butylphosphino)indol ≥98%
N-Phenyl-2-(di-t-butylphosphino)indol

cataCXium, Phosphine

Formula: C₂₂H₂₈NP
MW: 337.44 g/mol
MDL Number: MFCD06798302
CAS Number: 740815-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR