Octadecylphosphonic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-3510-1G
15-3510-5G
101418-660EA
103.32
USD
101418-660
101442-428
Octadecylphosphonic acid ≥97%
Octadecylphosphonic acid
Formula:
C₁₈H₃₉O₃P Melting Pt: 100…101 °C |
MDL Number:
MFCD00015838 CAS Number: 4724-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...