N-(2-Methoxyphenyl)-2-(di-t-butylphosphino)pyrrole ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

15-2975-2G 15-2975-500MG
100206-380EA 460 USD
100206-380 100198-646
N-(2-Methoxyphenyl)-2-(di-t-butylphosphino)pyrrole ≥95%
N-(2-Methoxyphenyl)-2-(di-t-butylphosphino)pyrrole

cataCXium, Phosphine

MDL Number: MFCD06798301
CAS Number: 1053658-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR