(-)-DIOP ≥99.5%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-DIOP, (4R,5R)-(-)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (((4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)

15-2970-250MG 15-2970-1G
100198-644EA 83.51 USD
100198-644 100196-094
(-)-DIOP ≥99.5%
(-)-DIOP

Phosphine

Formula: C₃₁H₃₂O₂P₂
MW: 498.532 g/mol
Melting Pt: 87…91 °C
MDL Number: MFCD00014107
CAS Number: 32305-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR