(+)-DIOP ≥99.5%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (4S,5S)-(+)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (S,S)-DIOP, (((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene))bis(diphenylphosphine)

15-2960-1G 15-2960-250MG
100207-350EA 200.98 USD
100207-350 100207-584
(+)-DIOP ≥99.5%
(+)-DIOP

Phosphine

Formula: C₃₁H₃₂O₂P₂
MW: 498.532 g/mol
Melting Pt: 87…91 °C
MDL Number: MFCD00009760
CAS Number: 37002-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR