Diphenyl(m-sulfonatophenyl)phosphine dihydrate sodium salt ≥90%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Diphenyl(m-sulphonatophenyl)phosphine dihydrate sodium salt

15-1827-5G 15-1827-1G
101324-624EA 962.46 USD
101324-624 101324-622
Diphenyl(m-sulfonatophenyl)phosphine dihydrate sodium salt ≥90%
Diphenyl(m-sulfonatophenyl)phosphine dihydrate sodium salt

Phosphine, Phosphine, Water Soluble

Formula: (C₆H₅)₂P(C₆H₄SO₃Na)·2H₂O
MW: 400.36 g/mol
MDL Number: MFCD07772021
CAS Number: 63995-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR