Chlorobis(2-methylphenyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Di-o-tolylchlorophosphine, Chlorodi(o-tolyl)phosphine

15-1815-5G 15-1815-1G
100208-322EA 399.14 USD
100208-322 100208-320
Chlorobis(2-methylphenyl)phosphine ≥98%
Chlorobis(2-methylphenyl)phosphine

Phosphine

Formula: C₁₄H₁₄ClP
MW: 248.688 g/mol
MDL Number: MFCD04038733
CAS Number: 36042-94-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR