2-Diphenylphosphino-6-methylpyridine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-1804-500MG
15-1804-2G
100198-642EA
158.52
USD
100198-642
100206-366
2-Diphenylphosphino-6-methylpyridine ≥98%
2-Diphenylphosphino-6-methylpyridine
Phosphine
Formula:
C₁₈H₁₆NP Melting Pt: 81…83 °C |
MDL Number:
MFCD04974234 CAS Number: 132682-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...