Chloro(diisopropyl)phosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Di-i-propylchlorophosphine

15-1800-5G 15-1800-1G
100201-482EA 287.32 USD
100201-482 100201-480
Chloro(diisopropyl)phosphine ≥97%
Chloro(diisopropyl)phosphine

Phosphine

Formula: C₆H₁₄ClP
MW: 152.6 g/mol
Density: 0.959
Flash Pt: 39 °F
MDL Number: MFCD00015027
CAS Number: 40244-90-4
UN: 2924
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR