(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-(-)-2-(Diphenylphosphino)-2'-methoxy-1,1'-binaphthyl, (S)-(2'-Methoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine, (S)-(-)-MOP, (S)-MOP
15-1776-500MG
15-1776-100MG
100199-802EA
560.49
USD
100199-802
100199-800
(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥99%
(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl
Phosphine
Formula:
C₃₃H₂₅OP MW: 468.536 g/mol Melting Pt: 175…179 °C |
MDL Number:
MFCD00269686 CAS Number: 134484-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...