2-(Diphenylphosphino)ethylamine ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

15-1762-5G 15-1762-1G
101418-634EA 963.88 USD
101418-634 101418-632
2-(Diphenylphosphino)ethylamine ≥95%
2-(Diphenylphosphino)ethylamine

Phosphine

Formula: (C₆H₅)₂PCH₂CH₂NH₂
MDL Number: MFCD00233831
CAS Number: 4848-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR