2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Diphenylphosphino-2'-(N,N-dimethylamino)biphenyl, PhDavePhos, 2-Diphenylphosphino-2'-(N,N-dimethylamino))-1,1'-biphenyl

15-1745-2G 15-1745-500MG
100198-014EA 142.95 USD
100198-014 100206-364
2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl ≥98%
2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₆H₂₄NP
MDL Number: MFCD03094578
CAS Number: 240417-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR