Diphenylphosphinic chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Diphenylphosphinyl chloride

15-1740-25G 15-1740-5G 15-1740-100G
100196-086EA 208.06 USD
100196-086 100209-716 100207-348
Diphenylphosphinic chloride ≥98%
Diphenylphosphinic chloride

Phosphine

Formula: C₁₂H₁₀ClOP
MW: 236.63 g/mol
Boiling Pt: 138…139 °C (0.15 mmHg)
Density: 1.240
Flash Pt: 79 °F
MDL Number: MFCD00002077
CAS Number: 1499-21-4
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR