(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)[(1R)-1-phenylethyl]amine ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-N-(1-Phenylethyl)dinaphtho[2,1-d:1,2-f][1,3,2]dioxaphosphepin-4-amine, (S,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)-(1-phenylethyl)amine, (+)-N-(1(R1-Phenylethyl)-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine,.(11b,S)

15-1525-100MG 15-1525-500MG
100201-476EA 138.71 USD
100201-476 100201-478
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)[(1R)-1-phenylethyl]amine ≥95%
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)[(1R)-1-phenylethyl]amine

MONOPHOS, Phosphine

Formula: C₂₈H₂₂NO₂P
MW: 435.45 g/mol
Melting Pt: 212…213 °C
MDL Number: MFCD04973053
CAS Number: 422509-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR