Di-2-norbornylphosphine 10% (w/w) in hexanes ≥98% (mixture of endo and exo isomers)

Supplier: STREM CHEMICALS INC MS
Danger

15-1461-50G 15-1461-10G
100211-018EA 400.55 USD
100211-018 100201-470
Di-2-norbornylphosphine 10% (w/w) in hexanes ≥98% (mixture of endo and exo isomers)
Di-2-norbornylphosphine

Phosphine

MDL Number: MFCD04117684
CAS Number: 148432-44-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR