Di-2-norbornylphosphine ≥98% (mixture of endo and exo isomers)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
15-1460-5G
15-1460-1G
100207-576EA
270.34
USD
100207-576
100207-574
Di-2-norbornylphosphine ≥98% (mixture of endo and exo isomers)
Di-2-norbornylphosphine
Phosphine
Density:
0,88 |
MDL Number:
MFCD04117684 CAS Number: 148432-44-4 UN: 2845 ADR: 4.2,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...