(S)-(+)-(2,6-Dimethyl-3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (11bS)-N,N,2,6-Tetramethyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

15-1255-500MG 15-1255-100MG
100197-072EA 1125.23 USD
100197-072 100197-070
(S)-(+)-(2,6-Dimethyl-3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98%
(S)-(+)-(2,6-Dimethyl-3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine

MONOPHOS, Phosphine

Formula: C₂₄H₂₂NO₂P
MW: 387.4100 g/mol
Melting Pt: 228…229 °C
MDL Number: MFCD04117683
CAS Number: 185449-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR