4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 9,9-Dimethyl-4,5-bis(di-t-butylphosphino)xanthene, t-Bu-Xantphos, (9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(di-tert-butylphosphine)

15-1241-2G 15-1241-500MG
101324-610EA 539.26 USD
101324-610 101324-608
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene ≥97%
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene

Phosphine, Pincer Ligands and Complexes, XANTPHOS

Formula: C₃₁H₄₈OP₂
MW: 498.68 g/mol
MDL Number: MFCD07781995
CAS Number: 856405-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR