(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)morpholine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-4-(Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)morpholine, (S)-MorfPhos
15-1235-500MG
15-1235-100MG
101433-152EA
297.23
USD
101433-152
101433-150
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)morpholine ≥97%
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)morpholine
MONOPHOS, Phosphine
Formula:
C₂₄H₂₀NO₃P MW: 401.3900 g/mol |
MDL Number:
MFCD08276866 CAS Number: 185449-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...