Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Dicyclohexylphosphino-2’,6’-dimethoxybiphenyl, SPhos, Dicyclohexyl(2',6'-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl

15-1143-500MG 15-1143-2G
100200-426EA 55.2 USD
100200-426 100203-744
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98%
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₆H₃₅O₂P
MW: 410.53 g/mol
Melting Pt: 164…166 °C
MDL Number: MFCD05861611
CAS Number: 657408-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR