Dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphine

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cy-vBRIDP®, Dicyclohexyl(2,2-diphenyl-1-methylvinyl)phosphine

15-1062-250MG 15-1062-1G
101442-350EA 46.71 USD
101442-350 101442-354
Dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphine
Dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphine

Phosphine

Formula: C₂₇H₃₅P
MW: 390.541 g/mol
MDL Number: MFCD10565627
CAS Number: 384842-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR