(S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-Cl-MeO-BIPHEP

15-1056-250MG 15-1056-1G
100196-082EA 171.26 USD
100196-082 100196-084
(S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl ≥95%
(S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₃₈H₃₀Cl₂O₂P₂
Melting Pt: 178 °C
MDL Number: MFCD03426987
CAS Number: 185913-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR