(R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-(5,5'-Dichloro-6,6'-dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(diphenylphosphine), (R)-Cl-MeO-BIPHEP

15-1055-250MG 15-1055-1G
100196-078EA 171.26 USD
100196-078 100196-080
(R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl ≥95%
(R)-(+)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₃₈H₃₀Cl₂O₂P₂
Melting Pt: 178 °C
MDL Number: MFCD03426987
CAS Number: 185913-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR