2-(Di-tert-butylphosphino)-2'-methylbiphenyl ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Di-t-butylphosphino-2'-methyl)-1,1'-biphenyl, t-BuMePhos, Di-tert-butyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine

15-1049-2G 15-1049-50G 15-1049-500MG 15-1049-10G
100197-998EA 244.86 USD
100197-998 100198-000 100206-344 100197-996
2-(Di-tert-butylphosphino)-2'-methylbiphenyl ≥99%
2-(Di-tert-butylphosphino)-2'-methylbiphenyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₁H₂₉P
MW: 312.437 g/mol
MDL Number: MFCD03453047
CAS Number: 255837-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR