2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Di-t-butylphosphino-2'-(N,N-dimethylamino))-1,1'-biphenyl, tBuDavePhos, 2'-(Di-tert-butylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine

15-1048-2G 15-1048-500MG
100199-782EA 226.46 USD
100199-782 100211-014
2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl ≥98%
2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₂H₃₂NP
MDL Number: MFCD03426986
CAS Number: 224311-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR