2-(Di-tert-butylphosphino)biphenyl ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-(Di-tert-Butylphosphino)biphenyl, JohnPhos, 2-(Di-t-butylphosphino))-1,1'-biphenyl

15-1045-2G 15-1045-500MG 15-1045-50G 15-1045-10G
100199-780EA 124.55 USD
100199-780 100211-012 100202-980 100202-978
2-(Di-tert-butylphosphino)biphenyl ≥99%
2-(Di-tert-butylphosphino)biphenyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₀H₂₇P
MW: 298.40 g/mol
Melting Pt: 85 °C
MDL Number: MFCD01862440
CAS Number: 224311-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR