racemic-2-Di-t-butylphosphino-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: TrixiePhos, [1,1'-Binaphthalen]-2-yldi-tert-butylphosphine

15-1043-1G 15-1043-250MG
100196-914EA 283.08 USD
100196-914 100200-868
racemic-2-Di-t-butylphosphino-1,1'-binaphthyl ≥98%
racemic-2-Di-t-butylphosphino-1,1'-binaphthyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₈H₃₁P
MW: 398.52 g/mol
Melting Pt: 147…149 °C
MDL Number: MFCD03094576
CAS Number: 255836-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR