Di-tert-butylphosphine 10% (w/w) in hexanes ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-1042-50G 15-1042-250G
100200-420EA 365.17 USD
100200-420 100197-068
Di-tert-butylphosphine 10% (w/w) in hexanes ≥98%
Di-tert-butylphosphine

Phosphine

Formula: C₈H₁₉P
MDL Number: MFCD00134241
CAS Number: 819-19-2
UN: 1993
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR