Di-t-butylphosphine oxide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-1041-250MG 15-1041-1G
100211-010EA 108.98 USD
100211-010 100199-778
Di-t-butylphosphine oxide ≥98%
Di-t-butylphosphine oxide
Formula: C₈H₁₉OP
MDL Number: MFCD00460156
CAS Number: 684-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR