Di-t-butylmethylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-1020-1G 15-1020-5G
100200-864EA 150.03 USD
100200-864 100200-866
Di-t-butylmethylphosphine ≥98%
Di-t-butylmethylphosphine

Phosphine

Density: 0,824
MDL Number: MFCD11869304
CAS Number: 6002-40-0
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR