Triphenylphosphine dibromide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dibromotriphenylphosphorane

15-0990-10G 15-0990-50G
100201-460EA 87.75 USD
100201-460 100207-346
Triphenylphosphine dibromide ≥98%
Triphenylphosphine dibromide

Phosphine

Formula: C₁₈H₁₅Br₂P
MW: 422.1 g/mol
MDL Number: MFCD00000054
CAS Number: 1034-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR