(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl) ≥94%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)-1-naphthamide), (R,R)-DACH-Naphthyl Trost Ligand
15-0963-100MG
15-0963-500MG
15-0963-2G
100198-624EA
62.28
USD
100198-624
100198-626
100199-772
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl) ≥94%
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl)
Phosphine, TROST
Formula:
C₅₂H₄₄N₂O₂P₂ MW: 790.8655 g/mol |
MDL Number:
MFCD02684551 CAS Number: 174810-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...