(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: N,N'-((1S,2S)-Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)benzamide), (S,S)-DACH-Phenyl Trost Ligand

15-0961-100MG 15-0961-500MG 15-0961-2G
100203-978EA 59.45 USD
100203-978 100203-980 100209-696
(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) ≥95%
(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl)

Phosphine, TROST

Formula: C₄₄H₄₀N₂O₂P₂
MW: 690.7481 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD01631274
CAS Number: 169689-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR