(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (1R,2R)-N,N'-(Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)benzamide), (R,R)-DACH-Phenyl Trost Ligand

15-0960-2G 15-0960-500MG 15-0960-100MG
100209-694EA 707.69 USD
100209-694 100203-976 100203-974
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) ≥98%
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl)

Phosphine, TROST

Formula: C₄₄H₄₀N₂O₂P₂
MW: 690.7481 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD01631273
CAS Number: 138517-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR