Decylphosphonic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
n-Decylphosphonic acid, 1-Decylphosphonic acid
15-0958-5G
15-0958-1G
101442-408EA
237.78
USD
101442-408
101442-406
Decylphosphonic acid ≥97%
Decylphosphonic acid
Formula:
C₁₀H₂₃O₃P MW: 222.26 g/mol Melting Pt: 103…104 °C |
MDL Number:
MFCD00015830 CAS Number: 6874-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...