(S)-(+)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-(+)-4,12-Bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane

15-0731-500MG 15-0731-100MG
100207-564EA 627.02 USD
100207-564 100207-562
(S)-(+)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS ≥97%
(S)-(+)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS

PHANEPHOS, Phosphine

Formula: C₄₈H₅₀P₂
MW: 688.858 g/mol
MDL Number: MFCD03840578
CAS Number: 325168-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR