(R)-(-)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(R)-3,5-xylyl-PHANEPHOS ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-(-)-4,12-Bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane

15-0730-100MG 15-0730-500MG
100207-558EA 157.11 USD
100207-558 100207-560
(R)-(-)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(R)-3,5-xylyl-PHANEPHOS ≥97%
(R)-(-)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(R)-3,5-xylyl-PHANEPHOS

PHANEPHOS, Phosphine

Formula: C₄₈H₅₀P₂
MW: 688.858 g/mol
MDL Number: MFCD03840578
CAS Number: 325168-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR